Masterflex® Fitting, Nylon, Straight, Female Luer to Hose Barb Adapter, 5/32" ID; 25/PK
Catalog # MFLX45502-06
Supplier: Avantor Fluid Handling
CE certificate
Avantor exclusive
About this item
Verzeker uw procesintegriteit met volledige materiaaltraceerbaarheid.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance
- Connection Type: Female Luer x 5/32" ID
- Ideal for use with flexible tubing
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
- Weerhaakmaat/buis inw.Ø (poort 1):5/32"
- Verbindingstype:Female Luer to Barb
- Flow pattern:Straight
- Omschrijving:Fitting, Nylon, Straight, Female Luer to Hose Barb Adapter, 5/32" ID; 25/PK
- Seq:07
- Environmentally Preferable:
- Connection type (port 1):Female luer
- No. of ports:2
- Garantie:3 Months
- VPE:25
- Fitting material:Nylon
Frequently Bought Together