Masterflex® Fitting, Polycarbonate, Three-Way Stopcock, Male Luer Lock, Non-Sterile; 10/PK
Catalogus # MFLX30600-02
Leverancier: Avantor Fluid Handling
Over dit artikel
Reguleer de stroom zonder te lekken.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Three-way stopcock/flow diverter with male luer lock connections
- HDPE handle rotates 180°
Kies kranen met een mannelijke luer-lock of een mannelijke luer-slip. Handgreep draait 90° op eenwegkranen, 180° op driewegkranen en 360° op vierwegkranen. Ze hebben allemaal een pc-behuizing en een HDPE-stam.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
- Flow pattern:Three-Way
- Omschrijving:Fitting, Polycarbonate, Three-Way Stopcock, Male Luer Lock, Non-Sterile; 10/PK
- Environmentally Preferable:
- Wetted materials:Polycarbonate
- Garantie:Out of box
- VPE:10
- Inlet port:Female Luer
- Outlet port:Male Luer Lock
Frequently Bought Together