PendoTECH® Single-Use Pressure Sensor Accessories
Adapter Tee, 1/4" Hose Barb x 1/4" Hose Barb x Male Luer Fitting, Polycarbonate
Leverancier: Avantor Fluid Handling
PendoTECH® Single-Use Pressure Sensor Accessories
Catalogus # MFLX19406-34
Leverancier: Avantor Fluid Handling
Ga naar:
Product Details & Documents
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
Certificaten
Zoeken op lotnummerSpecificaties
- Omschrijving:Adapter Tee, 1/4" Hose Barb x 1/4" Hose Barb x Male Luer Fitting, Polycarbonate
- Cat. No.:MFLX19406-34
- Process connection:1/4" x 1/4" hose barb, male luer
- Garantie:3 Months
- Max. druk:76 psi