Masterflex® Remote Control Cable Connectors, Avantor®
DB25 Male connector; for creating your own cable
Catalogus # MFLX07523-94
Leverancier: Avantor Fluid Handling
Masterflex® Remote Control Cable Connectors, Avantor®
Catalogus # MFLX07523-94
Leverancier: Avantor Fluid Handling
Ga naar:
Product Details & Documents
Create your own cable for optimization of your Masterflex® drive.
- Choose the correct connection for your Masterflex® pump
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).
Documents
Masterflex LS MasterSense Drives Pump
Product documentatie
Specificaties
- Omschrijving:DB25 Male connector; for creating your own cable
- Cat. No.:MFLX07523-94
- Type bediening:Analog Variable Speed
- Model:07523-94
- Garantie:2 years