CPC® Miniature Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 1/8" ID, SMM0212; 10/PK
Catalog # MFLX06363-60
Supplier: Avantor Fluid Handling
About this item
No more kinked tubing - allows tubing to rotate freely.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Twist-to-connect couplings provide a reliable and more secure alternative to luer-type connections
- Tubing can rotate freely when connected - preventing kinked tubing and accidental disconnection
These CPC® miniature quick-disconnect couplings are able to handle ¹/₁₆" ID tubing, and offer a more secure connection than luer fittings. Fitting halves engage and seal with a twist. Once engaged, the coupling allows the tubing to rotate freely.
For a complete coupling, order one fitting body and one insert.
- Weerhaakmaat/buis inw.Ø (poort 1):⅛"
- Verbindingstype:Quick-Disconnect to Barb
- Flow pattern:Straight
- Maximumtemperatuur:82 °C (180 °F)
- Omschrijving:Miniature Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, ¹/₈" ID, SMM0212; 10/PK
- Environmentally Preferable:
- O-Ring material:EPDM
- Valve material:None
- Min. temperatuur:0 °C (32 °F)
- Sterilisatie:Ethylene oxide (ETO) or gamma
- Garantie:3 Months
- VPE:10
- Fitting material:Polypropylene
- Quick-disconnect flow size:³⁄₃₂"
Frequently Bought Together