Masterflex® Smart Gasket® Sanitary Sampling Gasket, Viton, 2 Port, 1-1/2" Tri-Clamp; 1/EA
Catalogus # MFLX30543-14
Leverancier: Avantor Fluid Handling
Over dit artikel
Smart Gasket lets you take samples or readings from a sanitary line.
- Measure temperature, take a sample, or dose chemicals into your process line
- Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
- Sanitary clamp size (Port 1):1-1/2"
- Connection type:Sanitary
- Omschrijving:Sanitary Sampling Gasket, Viton, 2 Port, 1-1/2" Tri-Clamp; 1/EA
- Seq:35
- Environmentally Preferable:
- VPE:1
- Fitting material:Viton®
- Sanitary gasket size:1-1/2"
- No. of ports:2
- Garantie:1 year
Frequently Bought Together