Masterflex® Sanitary Gasket, EPDM, 3/4" Tri-Clamp®; 10/PK
Catalog # MFLX30548-60
Supplier: Avantor Fluid Handling
CE certificate
Specifications
See all product configurations- Catalog No:
- MFLX30548-60
- MFLX30548-84
- MFLX29590-83
- MFLX30548-86
- MFLX30548-80
- MFLX30548-82
- MFLX30548-22
- MFLX30548-04
- MFLX16011-66
- MFLX30548-06
- MFLX30548-00
- MFLX30548-88
- MFLX30548-02
- MFLX29590-80
- MFLX41001-77
- MFLX00404-MO
- MFLX00364-YQ
- MFLX30548-66
- MFLX30548-68
- MFLX30548-62
- MFLX30548-64
- MFLX30548-40
- MFLX00162-NJ
- MFLX30548-42
- MFLX30548-26
- MFLX30548-48
- MFLX30548-28
- MFLX30548-44
- MFLX30548-46
- MFLX30548-20
- MFLX30548-24
- MFLX30548-08
- Connection type:
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Garantie:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- Currently Viewing
Specifications
About this item
Ensure a tight seal.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Single-use custom configuration solutions to minimise downtime
These gaskets are for Tri-Clamp® style fittings and come in a wide variety of materials to match specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton® gaskets are animal-derived ingredient free and meet USP Class VI and cytotoxicity criteria.
Platinum-cured silicone is ideal for most pharmaceutical, food, and beverage processing applications. Odorless and tasteless to ensure media purity. May undergo SIP. Usable from –58 to 446 °F (–50 to 230 °C).
Viton offers excellent chemical resistance, especially to many acids and alkalis.
- Sanitary clamp size (Port 1):3/4"
- Connection type:Sanitary
- Omschrijving:Sanitary Gasket, EPDM, 3/4" Tri-Clamp®; 10/PK
- Seq:07
- Environmentally Preferable:
- VPE:10
- Fitting material:EPDM
- Sanitary gasket size:3/4"
- Garantie:1 year
Frequently Bought Together