Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

CF® Dye Dextran 70000 MW, anionic and fixable

CF350-Dextran 70,000 MW, anionic and fixable, 347/448 nm, 1 mg
  80138.
 :   Biotium
CF® Dye Dextran 70000 MW, anionic and fixable
  80138.
 :  Biotium
 :  
CF® Dye Dextran 70000 MW, anionic and fixable
CF® Dye Dextran 70000 MW, anionic and fixable

CF350-Dextran 70,000 MW, anionic and fixable, 347/448 nm, 1 mg

 :  
 
190,00 €
   
1 * 1 mg
 

CF® dye labeled dextrans could be used as a fluorescent fluid-phase markers to study cell permeability, endocytosis, or mechanisms of biomolecular delivery. The dextran is 70,000 MW, and contains a formaldehyde-fixable free-amine group.

  • Water-soluble fluid phase markers for tracking bulk endocytosis and cell permeability
  • Available with molecular weights of 3,5 kD, 10 kD, 40 kD, 70kD, 150 kD, or 250 kD
  • Choose from a selection of bright and photostable CF® dyes

Dextrans are water soluble branched-chain polysaccharides. Fluorescently labeled dextrans are used as markers for trafficking of fluid phase endocytic cargo to lysosomes. They are also used as tracers for epithelial and endothelial permeability. Labeled dextrans may also be used as microinjected tracers for neuronal morphology. CF® Dye Dextrans are available with a variety of dye colors and molecular weights ranging from 3,500 to 250,000.

These conjugates are available with Biotium's next-generation CF® dyes, a series of highly water-soluble fluorescent dyes spanning the visible and near-infrared spectrum for labeling biomolecules such as proteins and nucleic acids. Developed in house by Biotium's industry-leading chemists, CF® dyes offer superior brightness, photostability, and signal-to-noise when compared to other commercially available fluorescent dyes.

 

 

  • VPE:
    1 mg
  • Omschrijving:
    CF350-Dextran 70,000 MW, anionic and fixable
  • Ex/Em:
    347/448 nm

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.