Mobile safety steps
Mobile safety steps, 3 steps and platform, 860×1135×1000 mm
Catalogus # TEKMMST01000-3
Leverancier: TEKNOMEK
Ga naar:
Product Details & Documents
Our mobile safety steps offer optimal durability and toughness, being fully welded in 304 grade stainless steel, offering a non-slip surface for maximum safety.
- Whole structure comes with a brushed dull polish finish
- Stainless steel non slip check plate steps to ensure maximum safety and security for workers
- 3 castors- 2 swivel to rear and 1 retractable fixed at front
- Locking handle
- Widened legs
The steps castors and swivel ensure ease of manoeuvre, and the locking handle secures the steps ready for use. Different heights are available.
Specificaties
- Omschrijving:Mobile safety steps, 3 steps and platform
- Seq:02
- b×d×h:860×1135×1000 mm
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.