Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Natriumhydroxide 35% in aqueous solution, GPR RECTAPUR®

Natriumhydroxide 35% in aqueous solution, GPR RECTAPUR®

Plastic drum, 200 L
Catalogus # 28224.550
Corrosief
Leverancier:   VWR Chemicals
Natriumhydroxide 35% in aqueous solution, GPR RECTAPUR®
Catalogus # 28224.550
Leverancier:  VWR Chemicals
CAS-nummer:  1310-73-2
Natriumhydroxide 35% in aqueous solution, GPR RECTAPUR®
Natriumhydroxide 35% in aqueous solution, GPR RECTAPUR®

Plastic drum, 200 L

CAS-nummer:  1310-73-2
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 200 L
Product Details & Documents
Synoniemen:  Caustic soda
  • CAS-nummer:1310-73-2
  • Formule:NaOH
  • MDL-nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Kokend Pt:119…135 °C (1013 hPa)
  • Smelten Pt:9…10 °C
  • Dichtheid:∼1,38 g/ml
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specificaties

Specificatie testresultaten

  • Assay:
    34 - 36 %
  • Density (20/4):
    1.370 - 1.390
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Fe (Iron):
    Max. 20 ppm

Specificaties

  • Verpakking:
    Plastic drum
  • VPE:
    200 L

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.