Orders LinkContactUsLinkComponent
Accessories for multi-use data loggers, tempmate.®-M2
Accessories for multi-use data loggers, tempmate.®-M2
External digital temperature and humidty sensor; 1,2 m length, –40...+90 °C, 0 - 100% RH
Catalogus # IMECM2S-200
Leverancier:   tempmate
Accessories for multi-use data loggers, tempmate.®-M2
Catalogus # IMECM2S-200
Leverancier:  tempmate
Accessories for multi-use data loggers, tempmate.®-M2
Accessories for multi-use data loggers, tempmate.®-M2
External digital temperature and humidty sensor; 1,2 m length, –40...+90 °C, 0 - 100% RH
Price
26,20 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
  • Omschrijving:
    External digital temperature and humidty sensor; 1,2 m length, –40...+90 °C, 0 - 100% RH
  • Seq:
    02