Watch glasses
Watch glass, glass, with fused rim, 100 mm
Catalogus # HECH40010010
Leverancier: GLASWARENFABRIK KARL HECHT
Ga naar:
Product Details & Documents
Evaporating dishes, made of glass, with fused rim.
- Good chemical restistance
- Available with or without hole
Certifications: DIN 12 341
Specificaties
- Ø:100 mm
- Omschrijving:Watch glass, glass, with fused rim
- Seq:16