[EN]CITIFLUOR CFM-3 MOUNTING MEDIUM 25ML 1 * 25 mL
SCIAE17979-30
Frequently Bought Together
5-Fluorouracil ≥99% (by HPLC), powder, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
[EN]1 COMPONENT OS 1000MG/L IN HCL 2% 1 * 100 mL
: CUSTOM MADE CHEMICALS LAB
Photometric test kits Robot tests
: MACHEREY-NAGEL
[EN]POTASSIUM CYANIDE SOLUTION 1 * 100 mL
: PROLABO CMP
Kaliumhydroxide ≥85% ACS (max. 2% K₂CO₃)
: Thermo Fisher Scientific
[EN]65ML PL SILICA LR 2 (162 TESTS) 1 * 65 mL
: WATER ID PRIMELAB
Test strips, water hardness, AQUADUR®
: MACHEREY-NAGEL
[EN]50 TABLETS MOLYBDATE LR N 2 PHOTOME 1 * 50 ST
: WATER ID PRIMELAB