Orders LinkContactUsLinkComponent
Corning® 100-Layer CellCube® Module, Corning
Corning® 100-Layer CellCube® Module, Corning
100-layer CellCube® module, polycarbonate/polystyrene, sterile, 85000 cm²
Catalogus # CORN3032
Leverancier:   Corning
Corning® 100-Layer CellCube® Module, Corning
Catalogus # CORN3032
Leverancier:  Corning
Corning® 100-Layer CellCube® Module, Corning
100-layer CellCube® module, polycarbonate/polystyrene, sterile, 85000 cm²
Price
3 510,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.
It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth. These CellCube Modules provide: a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates, 85 000cm² polystyrene growth surface optimally treated for the growth of attachment dependent cells, a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen, an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. Square Flask Shape.
Specificaties
  • Omschrijving:
    100-layer CellCube® module, polycarbonate/polystyrene, sterile
  • Seq:
    01
  • Cultuuroppervlakte:
    85000 cm²
  • Oppervlakte:
    Tissue culture treated
  • Cat. No.:
    CORN3032
  • Mediavolume:
    6000 ml