Natriumhydroxide solution 50%, Biotech Reagent, J.T.Baker®
Catalogus # 2622.9025PE
Leverancier: Avantor
CAS-nummer: 1310-73-2
Ga naar:
Product Details & Documents
Synoniemen: Caustic soda
- CAS-nummer:1310-73-2
- Formule:NaOH
- MDL-nummer:MFCD00003548
- Molecular Weight:40 g/mol
- UN:1823
- ADR:8,II
Specificaties
Specificatie testresultaten
- Assay (NaOH):49 - 51 %
- Appearance (clear, colorless liquid):passes test
- Potassium (K):max. 0.01 %
- Identification (Na):passes test
- Sodium Carbonate (Na2CO3):max. 1.0 %
- Trace Impurities (in ppm) Chlorides (as Cl):max. 5
- Trace Impurities (in ppm) Iron (Fe):max. 2
- Trace Impurities (in ppm) Heavy Metals (as Pb):max. 1
Specificaties
- Verpakking:PE
- VPE:25 L
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



