Orders LinkContactUsLinkComponent
Condensers for REVOLVE 3M microscope
Condensers for REVOLVE 3M microscope
LWUCD universele revolvercondensor met lange werkafstand - NA 0,55 - WD 27 mm
Catalog # 630-3335
Supplier:   DISCOVER ECHO
Condensers for REVOLVE 3M microscope
Catalog # 630-3335
Supplier:  DISCOVER ECHO
Condensers for REVOLVE 3M microscope
LWUCD universele revolvercondensor met lange werkafstand - NA 0,55 - WD 27 mm
Price
3 090,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Omschrijving:
    LWUCD universele revolvercondensor met lange werkafstand - NA 0,55 - WD 27 mm
  • Seq:
    02