Multiple affinity removal starter reagent kits
Multiple affinity removal buffer B, 1 L, For eluting, 1
Catalogus # AGLS5185-5988
Leverancier: AGILENT
Multiple affinity removal starter reagent kits
Catalogus # AGLS5185-5988
Leverancier: AGILENT
CAS-nummer:
Ga naar:
Product Details & Documents
Starter reagent kit for multiple affinity removal LC columns.
Levering: Includes: 2 bottles buffer A (1 L), 1 bottle buffer B (1 L), 2 packs of 25 spin filters, 1 pack of 25 spin concentrator.
Specificaties
Specificaties
- VPE:1
- Omschrijving:Multiple affinity removal buffer B, 1 L
- Details:For eluting