Vessel cannulation forceps
0,6 mm Ext.Ø tubing size, Inox, 115 mm, Angled
Catalogus # FSCI18406-11
Marketsource
Leverancier: FINE SCIENCE TOOLS
Ga naar:
Product Details & Documents
Specificaties
- Omschrijving:0,6 mm Ext.Ø tubing size
- Lengte:115 mm
- Materiaal:Inox
- Seq:02
- Tip shape:Angled