Politainers, b.safe
20,0 L, S60, 285×285×295 mm
Catalog # 590-0307
Supplier: Bohlender
Jump to:
Product Details & Documents
Foldable canister for the collection of liquid waste.
- Including screw cap
- Foldable canister
Folded to save space during delivery and storage, unfolds automatically during filling. UN-approval only if used with the appropriate carton box.
Ordering information: Please order additional accessories like the carton box or the adaptor for Waste Caps GL 45 separately.
Specifications
- Schroefdraad:S60
- Capaciteit:20,0 L
- Seq:03
- b×d×h:285×285×295 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



