Politainers, b.safe
5,0 L, 38×3, 181×181×195 mm
Catalog # 590-0305
Supplier: Bohlender
Jump to:
Product Details & Documents
Foldable canister for the collection of liquid waste.
- Including screw cap
- Foldable canister
Folded to save space during delivery and storage, unfolds automatically during filling. UN-approval only if used with the appropriate carton box.
: Please order additional accessories like the carton box or the adaptor for Waste Caps GL 45 separately.
Specifications
- Schroefdraad:38×3
- Capaciteit:5,0 L
- Seq:01
- b×d×h:181×181×195 mm



