Orders LinkContactUsLinkComponent
Thread adaptors, TYPE A, b.safe
Thread adaptors, TYPE A, b.safe
PP, Tri-Sure 2" (male), G 2"/BSP 2" (female)
Catalogus # 590-0268
Leverancier:   Bohlender
Thread adaptors, TYPE A, b.safe
Catalogus # 590-0268
Leverancier:  Bohlender
Thread adaptors, TYPE A, b.safe
Thread adaptors, TYPE A, b.safe
PP, Tri-Sure 2" (male), G 2"/BSP 2" (female)
Price
62,30 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
All adaptors of this type are supplied with a male thread, a female thread and recessed grips.
  • Stay flexible and independent
  • Recessed grips for an easy assembly
Your existing caps and waste caps do not fit with the flasks and canisters which you would like to use or vice versa? Simply use b.safe thread adaptors for a flexible combination of caps and waste caps independently from the thread on the flask and/or container.
Specificaties
  • Schroefdraad:
    Tri-Sure 2" (male), G 2"/BSP 2" (female)
  • Materiaal:
    PP
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.