Multiports, b.safe
Tubing connector, 8× capillary connector, 8× fittings for tubing Ext.Ø 1,6 mm, 8× fittings for tubing Ext.Ø 2,2 mm, 8× fittings for tubing Ext.Ø 3,2 mm, NPT ¹/₈"
Catalogus # 590-0185
Leverancier: Bohlender
Ga naar:
Product Details & Documents
Extension of the number of capillary and/or tubing connectors on a b.safe Waste Cap. With PFA Fittings for capillaries with Ext.Ø 1,6/2,2/3,2 mm (thread UNF ¹/₄"), 1 hose connector for tubing Int.Ø 5,5 to 8,0 mm (thread NPT ¹/₈").
- Excellent chemical resistance
- Available for a wide range of hose diameters
- The conical shape gets the connection tight and leak-free
Adaptor to enlarge the number of capillary and tubing connectors of a b.safe waste cap.
Specificaties
- Schroefdraad:NPT ¹/₈"
- Met:8× capillary connector, 8× fittings for tubing Ext.Ø 1,6 mm, 8× fittings for tubing Ext.Ø 2,2 mm, 8× fittings for tubing Ext.Ø 3,2 mm
- Omschrijving:Tubing connector
- Seq:08