2-Bromopyridine
Bottle, 50 g
Catalog # APOSOR23057-50G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:109-04-6
- Formula:C₅H₄BrN
- MDL Number:MFCD00006219
- Molecular Weight:158 g/mol
- Boiling Pt:192…194 °C
- Density:1,657 g/cm³ (25 °C)
- Flash Pt:54 °C
- Storage Temperature:Kamertemperatuur
- UN:2810
- ADR:6.1,II
Specifications
Specifications
- Verpakking:Bottle
- VPE:50 g



