Isopropyl-beta-D-thiogalactopyranoside EMPROVE EXPERT 1 * 100 g
Catalogus # 1.37064.0100
Leverancier: MERCK PRODUCTION CHEMICALS
Isopropyl-beta-D-thiogalactopyranoside EMPROVE EXPERT 1 * 100 g
Catalogus # 1.37064.0100
Leverancier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Leverancier: Merck
Ethanol, GPR RECTAPUR®
Leverancier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Leverancier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Aceton ≥99.5%, GPR RECTAPUR®
Leverancier: VWR Chemicals
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
Aceton, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Leverancier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
Xyleen ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Ethanol denatured, EMSURE® for analysis, Supelco®
Leverancier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Leverancier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals