ContactUsLinkComponent
Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgA1 AM130 is a chimeric monoclonal antibody combining the constant domains of the human IgA1 molecule with mouse variable regions. 1 * 1 mg
Catalogus # ACRBS1N-M164-1MG
Leverancier:  ACROBIOSYSTEMS
Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgA1 AM130 is a chimeric monoclonal antibody combining the constant domains of the human IgA1 molecule with mouse variable regions. 1 * 1 mg
Catalogus # ACRBS1N-M164-1MG
Leverancier:  ACROBIOSYSTEMS
Frequently Bought Together