0,00 €
Bestelling invoeren
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
Orders Link
ContactUsLinkComponent
Microscoopobjectieven
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan achromat phase, LWD, N.A. 0,65, 40×, 2,0 mm
630-3322
:
DISCOVER ECHO
Objectives for Rebel Microscopes
630-3322
:
DISCOVER ECHO
Objectives for Rebel Microscopes
Plan achromat phase, LWD, N.A. 0,65, 40×, 2,0 mm
895,00 €
1 * 1 ST
-
+
Discover Echco Hybrid Microscope
ECHO Rebel Microscope Product Range
ECHO Rebel Microscope Specification
Vergroting:
40×
Omschrijving:
Plan achromat phase, LWD, N.A. 0,65
Seq:
18
Werkafstand:
2,0 mm
Hybrid microscope (upright + inverted), ECHO Rebel
#: 630-3346
: DISCOVER ECHO