Orders LinkContactUsLinkComponent
PAPR systems, PowerCap® Infinity®
PAPR systems, PowerCap® Infinity®
TH3P R SL replacement filters
Catalogus # JSPACAU660-000-400
Leverancier:   JSP
PAPR systems, PowerCap® Infinity®
Catalogus # JSPACAU660-000-400
Leverancier:  JSP
PAPR systems, PowerCap® Infinity®
PAPR systems, PowerCap® Infinity®
TH3P R SL replacement filters
Price
49,60 €
Log in voor beschikbaarheid
1 * 10 PAIRE
Product Details & Documents
A fully integrated TH3 powered air respirator offering four of the highest levels of protection in one intuitive head-mounted unit. The system provides complete above the neck 4-in-1 protection; respiratory, head, eye, and face.
  • 4-in-1 protection
  • Super strong ABS shell
  • Superior all-day protection
  • Optical class 1 visor
  • High capacity HEPA filters
  • Polycarbonate face shield
  • Rubber skirt
PowerCap® Infinity® is the perfect choice for many industries where dust poses a great danger to health. PowerCap® Infinity® removes the need for face fit testing, ensuring that a high volume of clean air is delivered to the breathing zone. High capacity HEPA filters coupled with aerodynamically optimised pre-filter covers ensure PowerCap® Infinity® delivers 160 litres per minute of clean air to the user with maximum battery efficiency. Filters can be changed easily with the quick-change bayonet system.
The optical class 1 visor offers impact protection at speeds of up to 270 mph. The visor has a replaceable peel-off visor protector to prolong its life. Developed to seamlessly integrate with Sonis® helmet-mounted ear defenders.
Certificeringen: Respiratory protection: EN12941:1998 + A2:2008 TH3P, APF 40. Head protection: EN397. Eye and face protection: EN166.1.B. Hearing protection (Optional extra): EN352-3. Approval: AS/NZS 1716:2012, AS/NZS 1337.1, AS/NZS 1801.
Specificaties
  • Omschrijving:
    TH3P R SL replacement filters
  • Seq:
    02
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.