Natriumhydroxide, pellets NF, Multi-Compendial, J.T.Baker®
20 kg
: Avantor
Meets N.F. Requirements, Meets J.P. Chemical Specifications, Meets E.P. Chemical Specifications, Meets B.P. Chemical Specifications, GMP Manufactured Product
- GMP Manufactured Product
- Bulk Pharmaceutical Chemical
: For Manufacturing, processing or repackaging
: Caustic soda
- :1310-73-2
- :NaOH
- :MFCD00003548
- :40 g/mol
- :14,08627
- :1390 °C (1013 hPa)
- :323 °C
- :2,13 g/cm³ (20 °C)
- :215-185-5
- :8,II
- :Management of Change (MOC) category = R
- :Kamertemperatuur
- :1823
- :01-2119457892-27
Documents
- NF - Identification A (Sodium):Passes Test
- NF - Identification B (pH):≥ 11.0
- NF - Insoluble Substances and Organic Matter:Passes Test
- NF - Potassium (K):≤ 0.5 %
- NF - Carbonate (as Na₂CO₃):≤ 3.0 %
- NF - Total Alkali:95.0 - 100.5 %
- NF - Content of Sodium:54.0 - 59.8 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Carbonate (as Na₂CO₃):≤ 2.0 %
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Chloride (Cl):≤ 200 ppm
- EP/BP - Sulfate (SO₄):≤ 200 ppm
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Assay (NaOH):97.0 - 100.5 %
- JP - Assay:95.0 - 101.0 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤ 0.050 %
- JP - Potassium (K):Passes Test
- JP - Carbonate (as Na₂CO₃):≤ 2.0 %
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:≤2.0%
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.:Passes Test
- VPE:20 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



