Orders LinkContactUsLinkComponent
Universal reference RNAs
Universal reference RNAs
Universal mouse reference RNA, 2×200 µg, 2
Catalogus # AGLS740100
Leverancier:   AGILENT
Universal reference RNAs
Catalogus # AGLS740100
Leverancier:  AGILENT
CAS-nummer:  
Universal reference RNAs
Universal mouse reference RNA, 2×200 µg, 2
CAS-nummer:  
Price
1 007,00 €
Log in voor beschikbaarheid
1 * 2 ST
Product Details & Documents
High-quality total RNA for human, mouse, and rat microarray gene-expression profiling that acts as a consistent control for standard data set comparisons.
  • Comprised of 10 cell lines for human, 11 for mouse and 14 for rat giving broad gene coverage
  • UHRR selected as one of the two standards in the FDA led Microarray Quality Control (MAQC) project
The reference RNA is designed to be used as a reference for uarray gene-profiling experiments.
Levering: Includes RNase-free water, 1,5 ml.
Let op: For research use only. Not for use in diagnostic procedures.
Specificaties
Specificaties
  • VPE:
    2
  • Omschrijving:
    Universal mouse reference RNA, 2×200 µg
Specificaties
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.