CLEANING CONCENTRATE TICKOPUR TR 13/10 L 1 * 10 L
Catalog # BAND6018
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® Nano™, Digitale timer, twee kanalen
Supplier: VWR Collection
Kegs, wide neck, UN approved, with screw lid
Supplier: BURKLE
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Schrijfblok, zelfklevend, TexWrite® TexNotes
Supplier: Texwipe
VWR®, Nitrilhandschoenen
Supplier: VWR Collection
pH-teststrips, MQuant®, Supelco®
Supplier: Merck
Ammoniak-oplossing 20%, GPR RECTAPUR®
Supplier: VWR Chemicals
Blood lancets, VITREX® Steel
Supplier: Vitrex Medical A/S
Flessen met brede hals en schroefdop, Corning® Gosselin™
Supplier: Corning
VWR®, Transferpipetten, Hoge Prestaties
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandschoenen
Supplier: VWR Collection
Brilliant cresyl blue, dark green powder C.I. 51010
Supplier: MP Biomedicals
TRAY - SINGLE WAFER 3 H22-30-0615 1 * 100 ST
Supplier: ENTEGRIS