RackScan
RackScan s, code scanner for side reading of SBS racks and cryostorage boxes, accessory for the RackScan product family
Catalogus # EPPE6002000.014
Leverancier: EPPENDORF
Ga naar:
Product Details & Documents
The RackScan instrument can read up to 96 barcoded vials in parallel.
- Camera based instrument resulting in rapid imaging and decoding
- Comfortable and easy plug-in system
- Small footprint, ideal for benchtop (and integration)
The RackScan can be equipped with the 's' accessory.
Specificaties
- Omschrijving:RackScan s, code scanner for side reading of SBS racks and cryostorage boxes, accessory for the RackScan product family
- Seq:01