Orders LinkContactUsLinkComponent
RackScan
RackScan
RackScan b, scanner (bottom reading) for 2D-coded tubes in SBS racks and cryostorage boxes, includes software RackScan analyser
Catalogus # EPPE6002000.006
Leverancier:   EPPENDORF
RackScan
Catalogus # EPPE6002000.006
Leverancier:  EPPENDORF
RackScan
RackScan
RackScan b, scanner (bottom reading) for 2D-coded tubes in SBS racks and cryostorage boxes, includes software RackScan analyser
Price
10 720,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
The RackScan instrument can read up to 96 barcoded vials in parallel.
  • Camera based instrument resulting in rapid imaging and decoding
  • Comfortable and easy plug-in system
  • Small footprint, ideal for benchtop (and integration)
The RackScan can be equipped with the 's' accessory.
Specificaties
  • Omschrijving:
    RackScan b, scanner (bottom reading) for 2D-coded tubes in SBS racks and cryostorage boxes, includes software RackScan analyser
  • Seq:
    02
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.