ContactUsLinkComponent
EZ Dry 1 litre Moisture trap - Activated Molecular sieve sachets for maintaining low water content. For volumes up to 1000ml. Smalle st bottleneck: 17mm 1 * 10 ST
Catalogus # MSPPHR-0104-Z010
EZ Dry 1 litre Moisture trap - Activated Molecular sieve sachets for maintaining low water content. For volumes up to 1000ml. Smalle st bottleneck: 17mm 1 * 10 ST
Catalogus # MSPPHR-0104-Z010
Frequently Bought Together