ContactUsLinkComponent
SULFURIC ACID 50% V/V IN AQUEOUS SOL. 1 * 1 L
Catalogus # PRLS90084.290
Leverancier:  PROLABO CMP
SULFURIC ACID 50% V/V IN AQUEOUS SOL. 1 * 1 L
Catalogus # PRLS90084.290
Leverancier:  PROLABO CMP
Frequently Bought Together