ContactUsLinkComponent
Cynomolgus Rhesus macaque CD47, His Tag (CD7-C52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Ac cession F7A802-1). In the region Gln 19 - Glu 141, the AA sequence of Cynomolgus and Rhesus macaque CD47 are homologu 1 * 1 EA
Catalog # ACRBCD7-C52H1-50UG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque CD47, His Tag (CD7-C52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Ac cession F7A802-1). In the region Gln 19 - Glu 141, the AA sequence of Cynomolgus and Rhesus macaque CD47 are homologu 1 * 1 EA
Catalog # ACRBCD7-C52H1-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together