ContactUsLinkComponent
Human Cynomolgus Rhesus macaque CD28, Mouse IgG2a Fc Tag, low endotoxin (CD8-H52A5) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession P10747-1). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolg 1 * 1 EA
N° de catalogue ACRBCD8-H52A5100UG
Fournisseur:  ACRO Biosystems
Human Cynomolgus Rhesus macaque CD28, Mouse IgG2a Fc Tag, low endotoxin (CD8-H52A5) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession P10747-1). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolg 1 * 1 EA
N° de catalogue ACRBCD8-H52A5100UG
Fournisseur:  ACRO Biosystems
Frequently Bought Together