Orders LinkContactUsLinkComponent
Detectable volumetric spoon, blue
Detectable volumetric spoon, blue
Detectable volumetric spoon, 50,0 ml
Catalog # 300-0405
Supplier:   BURKLE
Detectable volumetric spoon, blue
Catalog # 300-0405
Supplier:  BURKLE
Detectable volumetric spoon, blue
Detectable volumetric spoon, blue
Detectable volumetric spoon, 50,0 ml
Price
238,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
These samplers are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.
  • PS detectable, blue
  • Individually packaged and sterilised
  • Can be detected by colour scanners, metal detectors or X-rays
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • With stripper edge
  • For single use
  • Clean room manufactured (clean room class 7)
Volumetric and dosing spoons can be used to measure small amounts of powder, granulates and liquids exactly and dose them precisely. For precise measuring, excess medium is stripped off the upper edge of the spoon. The content volume is engraved on the handle.
The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, thus allowing for good emptying of small remaining amounts. The flat base makes it possible to set the volumetric spoon down without spilling the content.
The disposable dosing spoon is suitable for use as a sample spoon for taking small sample quantities. Since the plastic volumetric spoon was developed for single use, there is no cleaning and the possibility of cross-contamination and impurities is excluded.
The dosing spoons and volumetric spoons are suitable for precise measuring for chemical and pharmaceutical applications, cosmetics, animal feed and the food & beverage industry.
Certifications: Complies with EU food requirements and FDA regulations.
Specifications
  • Capaciteit:
    50,0 ml
  • Omschrijving:
    Detectable volumetric spoon
  • Seq:
    08