Slide storing system
with feet, PMMA, 490x475x700 mm, 1300/drawer, 5200 overall
Catalogus # 631-1284
Leverancier: KARTELL
Ga naar:
Product Details & Documents
Made from high impact PMMA and steel, this slide storage system is designed for secure classification and storage of microscope slides.
- Lightweight and functional
- Durable steel frame for stability
- Semi-transparent, ice coloured drawers provide an attractive finish
- Transparent lids permit easy drawer viewing, while protecting slides from dust and foreign bodies
- System holds up to 1300 slides per drawer, 5200 slides in total
The system is composed of 4 sliding drawers, a closing cover, and 4 fixed support feet. As an alternative, the system is also available with 4 swivel castors for added mobility. Each single drawer can hold up to 13 slide supports and transparent cover plates (must be ordered separately). Slide supports are easily removed, and will accommodate up to 100 microscope slides. Each cavity is numbered. The complete storing system can hold up to 5200 slides.
Specificaties
- Aantal objectglaasjes:1300/drawer, 5200 overall
- Omschrijving:with feet
- Materiaal:PMMA
- Seq:02
- b×d×h:490x475x700 mm