Orders LinkContactUsLinkComponent
Vessel dilators, balanced instruments
Roestvrij staal, 145 mm, Smooth, Angled, Ø 0,2 mm
  FSCI18603-14
Marketsource
Vessel dilators, balanced instruments
  FSCI18603-14
Marketsource
Vessel dilators, balanced instruments
Vessel dilators, balanced instruments
Roestvrij staal, 145 mm, Smooth, Angled, Ø 0,2 mm
271,50 €
1 * 1 ST
  • Lengte:
    145 mm
  • Materiaal:
    Roestvrij staal
  • Seq:
    01
  • Tip stijl:
    Smooth
  • Tipgrootte:
    Ø 0,2 mm
  • Tip shape:
    Angled