[EN]ADHESIVE CRYSTALBOND 555 1 STICK 1 * 1 ST
Catalog # SCIAE50401-01
Supplier: SCI MUENCHEN
Frequently Bought Together
NANOCOLOR® Photometric Tube Test Kits for Nitrate
Supplier: MACHEREY-NAGEL
Giemsa's stain solution voor microscopie, Sigma-Aldrich®
Supplier: Merck
Kaliumhydroxide ≥85%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Elastin stain solution acc. Weigert, Sigma-Aldrich®
Supplier: Merck
NANOCOLOR® Photometric Tube Test Kits for COD
Supplier: MACHEREY-NAGEL
Qualitative test paper phosphatesmo MI for phosphatase in milk
Supplier: MACHEREY-NAGEL
Nickel Test Strips, Dosatest
Supplier: VWR Chemicals
Copper test strips, Dosatest
Supplier: VWR Chemicals
Teststrips, glucosetest, Reflectoquant®, Supelco®
Supplier: Merck
Kjeldahl-tabletten
Supplier: THOMPSON & CAPPER
Ammoniak-oplossing 25%, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Test strips, nitrite, Dosatest
Supplier: VWR Chemicals
GLYCEROL 87% 1 * 250 mL
Supplier: PROLABO CMP
Amino acid reagent
Supplier: Hach
PHASE C 1 * 2,5 L
Supplier: PROLABO CMP