ContactUsLinkComponent
POLYPROPYLE SCREWCAP, BLACK CLOSED FOR REAGENTUBE (GL18) BUTYL/DARK GREY, 48° SHORE A, 3MM, 100P] 1 * 1.000 ST
Catalogus # MSPPSV181296
POLYPROPYLE SCREWCAP, BLACK CLOSED FOR REAGENTUBE (GL18) BUTYL/DARK GREY, 48° SHORE A, 3MM, 100P] 1 * 1.000 ST
Catalogus # MSPPSV181296
Frequently Bought Together