POLYPROPYLE SCREWCAP, BLACK CLOSED FOR REAGENTUBE (GL18) BUTYL/DARK GREY, 48° SHORE A, 3MM, 100P] 1 * 1.000 ST
Catalogus # MSPPSV181296
Leverancier: MARKET SOURCE PART PROCESS
POLYPROPYLE SCREWCAP, BLACK CLOSED FOR REAGENTUBE (GL18) BUTYL/DARK GREY, 48° SHORE A, 3MM, 100P] 1 * 1.000 ST
Catalogus # MSPPSV181296
Leverancier: MARKET SOURCE PART PROCESS
Frequently Bought Together
VWR®, maatcilinders, hoge vorm, klasse B, PMP TPX
Leverancier: VWR Collection
Container, brede hals, vierkant, behandeld
Leverancier: Kautex
Screw caps, PP, black with PE foam insert
Leverancier: Kautex
Emmers met klikdeksels
Leverancier: JOKEY PLASTIK
VWR®, Universele filtertips
Leverancier: VWR Collection
VWR®, Pipettips, Universeel
Leverancier: VWR Collection
Flessen, brede hals, vierkant, met draad
Leverancier: Kautex
Low retention filter pipette tips, ZEROTIP®
Leverancier: GUANGZHOU JET BIOFILTRATION
Dispenser tips, PD-Tips II
Leverancier: Brand
VWR®, maatcilinders, hoge vorm, klasse B, PP
Leverancier: VWR Collection
VWR®, Flessen, brede hals, met schroefdop
Leverancier: VWR Collection
VWR®, Macrocuvetten
Leverancier: VWR Collection
Bags, zip seal, writing area, Minigrip®
Leverancier: Flexico
Assistent Brush for Conical Centrifuge Tubes
Leverancier: GLASWARENFABRIK KARL HECHT
VWR® High Performance, Flessen, brede hals met schroefdop
Leverancier: VWR Collection
VWR® Bottles, Wide Neck, Square, with Screw Caps
Leverancier: LAB3 OWN PRODUCTION
VWR® Reagensreservoirs
Leverancier: VWR Collection