RE-GEL KIT WITH .3 CC GEL. (QTY 5) QTY 5 - EACH FILLED WITH .3CC GEL A SUFFICIENT AMOUNT TO REGEL APPROXIMATELY 50 SMALL ANIMAL TRAN SMITTERS 1 * 1 Kit
Catalogus # MSPP276-0039-001
Leverancier: MARKET SOURCE PART PROCESS
Frequently Bought Together