Orders LinkContactUsLinkComponent
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan apochromat, N.A. 0,9, 60×, 0,26 mm
Catalog # ECHLRMAG9602
Supplier:   DISCOVER ECHO
Objectives for Rebel Microscopes
Catalog # ECHLRMAG9602
Supplier:  DISCOVER ECHO
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan apochromat, N.A. 0,9, 60×, 0,26 mm
Price
4 750,00 €
Log in for availability
1 * 1 ST
Specifications
  • Vergroting:
    60×
  • Omschrijving:
    Plan apochromat, N.A. 0,9
  • Seq:
    08
  • Werkafstand:
    0,26 mm