Objectives for Rebel Microscopes
Plan achromat oil, N.A. 1,25, 100×, 0,2 mm
Catalogus # ECHLRMAG1002
Leverancier: DISCOVER ECHO
Ga naar:
Product Details & Documents
Specificaties
- Vergroting:100×
- Omschrijving:Plan achromat oil, N.A. 1,25
- Seq:05
- Werkafstand:0,2 mm