Objectives for Rebel Microscopes
Plan achromat phase, N.A. 0,65, 40×, 0,4 mm
Catalogus # ECHLRMAG0403
Leverancier: DISCOVER ECHO
Ga naar:
Product Details & Documents
Specificaties
- Vergroting:40×
- Omschrijving:Plan achromat phase, N.A. 0,65
- Seq:10
- Werkafstand:0,4 mm