Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Dextrin, Soluble fiber

500 g/mol
Catalogus # TCIAD5658-500G
Leverancier:   TCI
Dextrin, Soluble fiber
Catalogus # TCIAD5658-500G
Leverancier:  TCI
CAS-nummer:  9004-53-9
Dextrin, Soluble fiber

500 g/mol

CAS-nummer:  9004-53-9
Price
48,40 €
Log in voor beschikbaarheid
1 * 500 g/mol
Product Details & Documents
Synoniemen:  Dextrin
  • CAS-nummer:9004-53-9
  • Formule:(C₆H₁₂O₆)n
  • MDL-nummer:MFCD00081554
  • Molecular Weight:180.16 g/mol
  • Merck index:14,02953
  • EINECS:232-675-4
Specificaties

Specificatie testresultaten

  • Appearance:
    White to Almost white powder to crystal
  • Solubility in hot Water:
    transparency
  • Drying loss:
    max. 10.000 %
  • Ignition residue (Sulfate):
    max. 0.5 %
  • Reducing suger:
    max. 8.0

Specificaties

  • VPE:
    500 g/mol

Specificaties