Orders LinkContactUsLinkComponent
Objectives for Rebel Microscopes
Plan achromat, N.A. 0,65, 40×, 0,4 mm
  ECHLRMAG0402
Objectives for Rebel Microscopes
  ECHLRMAG0402
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan achromat, N.A. 0,65, 40×, 0,4 mm
1 * 1 ST
  • Vergroting:
    40×
  • Omschrijving:
    Plan achromat, N.A. 0,65
  • Seq:
    09
  • Werkafstand:
    0,4 mm