Labels, hazard warning
Hazard labels: drop, PIC W008-TRI 025-PE-ROLL/1, Polyester, zwart op geel, 25×18 mm, 250 labels/roll
Catalogus # BRDY827498
Leverancier: Brady
Ga naar:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specificaties
- Kleur:zwart op geel
- Omschrijving:Hazard labels: drop
- Afmetingen:25×18 mm
- Materiaal:Polyester
- Seq:04
- Model:PIC W008-TRI 025-PE-ROLL/1
- Verpakt:250 labels/roll