Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Electronic crimping tools

Electronic crimping tools

Electronic crimper for 13 mm aluminium crimp caps
  MANA735513
Electronic crimping tools
  MANA735513
Electronic crimping tools
Electronic crimping tools

Electronic crimper for 13 mm aluminium crimp caps

 
1 050,00 €
   
1 * 1 ST
 

These battery-powered electronic crimping tools are suitable for aluminium crimp and Flip Top / Flip Off caps (not suitable for 20 mm magnetic / bi-metal crimp caps).

  • Mobile tools for consistent and reproducible crimping results
  • Crimping pressure adjustable by the control unit on top of the tool
  • Long lasting lithium ion cell batteries (full battery charge for several hundred vials, life time of battery >1500 charges)

Certifications: CE certificate of conformity along with one year warranty.

 
  • Omschrijving:
    Electronic crimper for 13 mm aluminium crimp caps
  • Seq:
    02
Accessories

 

  •  
    Omschrijving
     
     
  • MANA735509
    Rack for 2 electronic crimping tools
       
    1 * 1 ST - 21,50 €

 

  •  
    Omschrijving
     
     
  • MANA735500
    Replacement battery 6,6 V, 8,6 Wh
       
    1 * 1 ST - 154,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.