Toebehoren
Inner chamber, reinforced, max. total load 350 kg, BF series
Catalog # BIND8012-1829
Supplier: Binder
Jump to:
Product Details & Documents
Specifications
- Omschrijving:Inner chamber, reinforced, max. total load 350 kg
- Seq:43
- Voor:BF series